| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000462113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018174.hg.1 |
ENST00000462113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000433640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000462113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018174.hg.1 |
ENST00000462113.1 |
Ensembl |