Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010267.hg.1 |
TC02000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010267.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010267.hg.1 |
ENST00000376700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010267.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010266.hg.1 |
TC02000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010266.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010266.hg.1 |
ENST00000376700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010266.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010268.hg.1 |
TC02000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010268.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010268.hg.1 |
ENST00000376700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010268.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161331.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161331.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161331.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161331.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161331.hg.1 |
TC0200008795.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200161332.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161332.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161332.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161332.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161332.hg.1 |
TC0200008795.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008795.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161333.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161333.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161333.hg.1 |
ENST00000329106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161333.hg.1 |
NM_001008743.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161333.hg.1 |
TC0200008795.hg.1 |
Clariom D TC |