Assay Details
| Gene Symbol: SP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103857.1 | 1 - 2 | 62 | 66 | ||
| GenBank mRNA | BM804969.1 | - | 1 - 2 | 48 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | TC1700012429.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206718.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | TC1700012429.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206714.hg.1 | NR_103857.1 | Refseq |

SP2-AS1
Genomic Map