| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171821.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171821.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171821.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171821.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171821.hg.1 |
TC0900010509.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171819.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171819.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171819.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171819.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171819.hg.1 |
TC0900010509.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171820.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171820.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171820.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171820.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171820.hg.1 |
TC0900010509.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
ENST00000566873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010509.hg.1 |
NR_109771.1 |
Refseq |