Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013377.3 | NP_037509.3 | 1 - 2 | 273 | 111 | |
| GenBank mRNA | AK091096.1 | - | 1 - 2 | 240 | 111 | |
| AK294431.1 | - | 1 - 2 | 329 | 111 | ||
| BC093744.1 | - | 1 - 2 | 122 | 7939589 | 111 | |
| BC093746.2 | - | 1 - 2 | 122 | 7939591 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000313.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294431.1,BC093744.1,BC093746.2,NM_013377.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | ENST00000402685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | ENST00000539469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003992.hg.1 | TC12000313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003988.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003988.hg.1 | ENST00000539469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003988.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003988.hg.1 | TC12000313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003989.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003989.hg.1 | ENST00000539469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003989.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003989.hg.1 | TC12000313.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151856.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151858.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151861.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151857.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |

PDZRN4
Genomic Map