| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039415.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039415.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039415.hg.1 |
TC01002532.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039416.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039416.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039416.hg.1 |
TC01002532.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013764.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202843.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202843.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202843.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202843.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202843.hg.1 |
TC0100013764.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202842.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202842.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202842.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202842.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202842.hg.1 |
TC0100013764.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202848.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202848.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202848.hg.1 |
ENST00000431311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202848.hg.1 |
NR_038928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202848.hg.1 |
TC0100013764.hg.1 |
Clariom D TC |