| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060845.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060845.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060845.hg.1 |
TC01004028.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060846.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060846.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060846.hg.1 |
TC01004028.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017952.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235217.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235217.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235217.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235217.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235217.hg.1 |
TC0100017952.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100235216.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235216.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235216.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235216.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235216.hg.1 |
TC0100017952.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100235215.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235215.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235215.hg.1 |
ENST00000357246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100235215.hg.1 |
NM_001004343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100235215.hg.1 |
TC0100017952.hg.1 |
Clariom D TC |