Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127364.2 | NP_001120836.1 | 4 - 5 | 744 | 116 | |
| NM_001127365.2 | NP_001120837.1 | 3 - 4 | 570 | 116 | ||
| XM_017012134.1 | XP_016867623.1 | 4 - 5 | 739 | 116 | ||
| XM_017012135.1 | XP_016867624.1 | 4 - 5 | 884 | 116 | ||
| GenBank mRNA | BC026000.1 | - | 3 - 4 | 502 | 116 | |
| BC042034.1 | - | 4 - 5 | 725 | 116 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |

FAM221A
Genomic Map