Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300932.1 | NP_001287861.1 | 3 - 4 | 568 | 76 | |
| NM_199136.4 | NP_954587.2 | 4 - 5 | 742 | 76 | ||
| XM_011515369.2 | XP_011513671.1 | 4 - 5 | 737 | 76 | ||
| XM_011515370.2 | XP_011513672.1 | 4 - 5 | 737 | 76 | ||
| XM_011515372.2 | XP_011513674.1 | 4 - 5 | 882 | 76 | ||
| GenBank mRNA | BC063130.1 | - | 4 - 5 | 656 | 30382529 | 76 |
| BU958830.1 | - | 3 - 4 | 500 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000150.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07002349.hg.1 | ENST00000344962.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002349.hg.1 | ENST00000409653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002349.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002349.hg.1 | TC07000150.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147954.hg.1 | TC0700006931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000344962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000409994.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000429719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000446234.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000462546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | ENST00000483090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127364.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001127365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_001300932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006931.hg.1 | NM_199136.4 | Refseq |

FAM221A
Genomic Map