Assay Details
| Gene Symbol: WIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080529.2 | NP_001073998.2 | 4 - 5 | 537 | 120 | |
| XM_017012522.1 | XP_016868011.1 | 3 - 4 | 545 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000188.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
actin cortical patch assembly
endocytosis
multicellular organism development
spermatogenesis
regulation of cell shape
actin filament-based movement
cell differentiation
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of actin nucleation
actin cortical patch localization
SH3 domain binding
actin filament binding
actin cortical patch assembly
endocytosis
multicellular organism development
spermatogenesis
regulation of cell shape
actin filament-based movement
cell differentiation
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of actin nucleation
actin cortical patch localization
SH3 domain binding
actin filament binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002942.hg.1 | ENST00000242140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002942.hg.1 | ENST00000409123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002942.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002942.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002942.hg.1 | TC07000188.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149057.hg.1 | TC0700007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000242140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | ENST00000409290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007061.hg.1 | NM_001080529.2 | Refseq |

WIPF3
Genomic Map