| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010404.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000603156.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174027.hg.1 |
TC0700010404.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
ENST00000605357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
NR_110119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
NR_110120.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174026.hg.1 |
TC0700010404.hg.1 |
Clariom D TC |