| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018758.hg.1 |
ENST00000227256.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018758.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018758.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018758.hg.1 |
TC11001488.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018759.hg.1 |
ENST00000227256.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018759.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018759.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018759.hg.1 |
TC11001488.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178421.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100178421.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178421.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100178421.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178421.hg.1 |
TC1100010316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010316.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178422.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100178422.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178422.hg.1 |
ENST00000524983.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100178422.hg.1 |
NM_001029865.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100178422.hg.1 |
TC1100010316.hg.1 |
Clariom D TC |