Assay Details
| Gene Symbol: LINC01234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110026.1 | 2 - 3 | 204 | 63 | ||
| GenBank mRNA | DB172006.1 | - | 2 - 3 | 204 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193478.hg.1 | TC1200012000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193480.hg.1 | TC1200012000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193486.hg.1 | TC1200012000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000510694.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000550905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | ENST00000551982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012000.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000547963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | ENST00000550223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | NR_110025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | NR_110026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193479.hg.1 | TC1200012000.hg.1 | Clariom D TC |

LINC01234
Genomic Map