Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259328.4 | XP_005259385.1 | 1 - 2 | 177 | 157 | |
| XM_005259329.3 | XP_005259386.1 | 1 - 2 | 177 | 157 | ||
| XM_017027381.1 | XP_016882870.1 | 1 - 2 | 173 | 157 | ||
| GenBank mRNA | DC341389.1 | - | 1 - 2 | 186 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259328.4, XM_005259329.3, XM_017027381.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |

ZNF527
Genomic Map