Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032453.1 | NP_115829.1 | 3 - 4 | 86 | ||
| GenBank mRNA | AK301135.1 | - | 3 - 4 | 269 | 86 | |
| BC014325.1 | - | 3 - 4 | 86 | |||
| DC341389.1 | - | 3 - 4 | 86 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000515.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19006843.hg.1 | ENST00000356178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006843.hg.1 | ENST00000483919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006843.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006843.hg.1 | TC19000515.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000588911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | ENST00000589615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007973.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | NM_032453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | TC1900007973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000356178.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000436120.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000483919.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000587349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | ENST00000588512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159607.hg.1 | NM_032453.1 | Refseq |

ZNF527
Genomic Map