Assay Details
| Gene Symbol: TOPORS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033991.1 | 1 - 2 | 552 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000129.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001480.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001480.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001480.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001480.hg.1 | TC09000129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001478.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001478.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001478.hg.1 | TC09000129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001482.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001482.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001482.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001482.hg.1 | TC09000129.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147132.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147129.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147123.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147124.hg.1 | TC0900006945.hg.1 | Clariom D TC |

TOPORS-AS1
Genomic Map