Assay Details
| Gene Symbol: TOPORS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033991.1 | 2 - 3 | 797 | 94 | ||
| NR_033992.2 | 2 - 3 | 338 | 94 | |||
| GenBank mRNA | CB048380.1 | - | 1 - 2 | 148 | 94 | |
| HY032383.1 | - | 2 - 3 | 339 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000129.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P136641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564403_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 78383_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 221979_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001479.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001479.hg.1 | TC09000129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001483.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001483.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001483.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09001483.hg.1 | TC09000129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147126.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147126.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147126.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147134.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000425533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000451672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000453396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000458036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | ENST00000540066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006945.hg.1 | NR_102377.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147127.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147127.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147127.hg.1 | ENST00000450093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147127.hg.1 | NR_033992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147127.hg.1 | TC0900006945.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 78383_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 78383_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221979_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221979_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221979_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221979_at | - | - |
| Human Genome U133A 2.0 Array | 78383_at | - | - |
| Human Genome U133A 2.0 Array | 78383_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at12 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at13 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at14 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221979_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at16 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 78383_at15 | - | - |
| Human Genome U95 Set | 71878_at | - | - |
| Human Genome U95 Set | 71878_PM_at | - | - |
| Human Genome U95 Set | 78383_at | - | - |
| Human Genome U95 Set | 78383_PM_at | - | - |
| Human Genome U133 Set | 78383_at | - | - |
| Human Genome U133 Set | 221979_at | - | - |
| Human Genome U133 Set | 78383_PM_at | - | - |
| Human Genome U133 Set | 221979_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038469 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 171810 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3253850 | NR_033991.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3253850 | NR_033992.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3253850 | CR604036.1 | human_transcriptome |

TOPORS-AS1
Genomic Map