| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008199.hg.1 |
ENST00000370602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008199.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008199.hg.1 |
TC10000704.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000423054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000423054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000423054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000423054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008591.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
ENST00000370602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
ENST00000596005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
NM_001010917.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158618.hg.1 |
TC1000008591.hg.1 |
Clariom D TC |