Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005247167.1 | XP_005247224.1 | 1 - 2 | 351 | 126 | |
| XM_011512490.2 | XP_011510792.1 | 1 - 2 | 351 | 126 | ||
| XM_011512491.2 | XP_011510793.1 | 1 - 2 | 351 | 126 | ||
| XM_011512492.2 | XP_011510794.1 | 1 - 2 | 351 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005247167.1, XM_011512490.2, XM_011512491.2, XM_011512492.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |

KBTBD12
Genomic Map