Assay Details
| Gene Symbol: ERICH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | HY026606.1 | - | 3 - 4 | 376 | 64 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000520816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000522092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000524139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000577187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | ENST00000578889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012253.hg.1 | NR_073397.1 | Refseq |

ERICH1-AS1
Genomic Map