Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242757.1 | NP_001229686.1 | 4 - 5 | 873 | 96 | |
| XM_011522897.2 | XP_011521199.1 | 5 - 6 | 921 | 96 | ||
| XM_011522899.2 | XP_011521201.1 | 5 - 6 | 858 | 96 | ||
| XM_017022962.1 | XP_016878451.1 | 2 - 3 | 523 | 96 | ||
| XM_017022963.1 | XP_016878452.1 | 2 - 3 | 2271 | 96 | ||
| XM_017022964.1 | XP_016878453.1 | 3 - 4 | 1427 | 96 | ||
| XM_017022965.1 | XP_016878454.1 | 2 - 3 | 2212 | 96 | ||
| XM_017022966.1 | XP_016878455.1 | 3 - 4 | 2050 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001349.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018694.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018694.hg.1 | TC16001349.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018695.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018695.hg.1 | TC16001349.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000568161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000603735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011257.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | TC1600011257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187693.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | NM_001242757.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | TC1600011257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000562855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000562916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000563595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | ENST00000614943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187692.hg.1 | NM_001242757.1 | Refseq |

SLC22A31
Genomic Map