| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020242.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020242.hg.1 |
TC03001172.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020244.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020244.hg.1 |
TC03001172.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013942.hg.1 |
ENST00000447550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013942.hg.1 |
ENST00000447550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020243.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020243.hg.1 |
TC03001172.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020241.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020241.hg.1 |
TC03001172.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300013942.hg.1 |
ENST00000447550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013942.hg.1 |
ENST00000447550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206691.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206692.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206694.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206694.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206694.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206694.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206694.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206695.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013941.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206690.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206690.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206690.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
ENST00000561645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
ENST00000564146.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
NM_001195279.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206693.hg.1 |
TC0300013941.hg.1 |
Clariom D TC |