Assay Details
| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303482.1 | NP_001290411.1 | 2 - 3 | 400 | 95 | |
| XM_011525928.1 | XP_011524230.1 | 4 - 5 | 6710 | 95 | ||
| GenBank mRNA | BC037871.2 | - | 2 - 3 | 379 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800162131.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800162131.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800162131.hg.1 | TC1800009058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800162131.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800162131.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |

SMIM21
Genomic Map