Assay Details
| Gene Symbol: LOC283194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033853.2 | 1 - 2 | 80 | 63 | ||
| GenBank mRNA | AK091446.1 | - | 1 - 2 | 80 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001816.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020987.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020987.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020987.hg.1 | TC11001816.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020986.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020986.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020986.hg.1 | TC11001816.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000525714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010955.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182691.hg.1 | TC1100010955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000532098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | ENST00000533954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | NR_033853.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182693.hg.1 | TC1100010955.hg.1 | Clariom D TC |

LOC283194
Genomic Map