Assay Details
| Gene Symbol: CASP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001226.3 | NP_001217.2 | 3 - 4 | 313 | 124 | |
| GenBank mRNA | BC000305.2 | - | 3 - 4 | 261 | 2820468 | 124 |
| BC004460.1 | - | 3 - 4 | 261 | 2820468 | 124 | |
| U20536.1 | - | 3 - 4 | 313 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001464.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
cellular component disassembly involved in execution phase of apoptosis
epithelial cell differentiation
regulation of apoptotic process
cysteine-type endopeptidase activity
protein binding
cysteine-type peptidase activity
identical protein binding
cysteine-type endopeptidase activity involved in apoptotic process
proteolysis
apoptotic process
cellular component disassembly involved in execution phase of apoptosis
epithelial cell differentiation
regulation of apoptotic process
cysteine-type endopeptidase activity
protein binding
cysteine-type peptidase activity
identical protein binding
cysteine-type endopeptidase activity involved in apoptotic process
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04020806.hg.1 | ENST00000265164.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04020806.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04020806.hg.1 | TC04001464.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400179545.hg.1 | TC0400011578.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000265164.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000352981.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000503684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000505486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000507550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000508203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | ENST00000510324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_001226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011578.hg.1 | NM_032992.2 | Refseq |

CASP6
Genomic Map