Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000429.2 | NP_000420.1 | 4 - 5 | 655 | 86 | |
| XM_005269842.4 | XP_005269899.1 | 5 - 6 | 885 | 86 | ||
| XM_005269843.4 | XP_005269900.1 | 4 - 5 | 774 | 86 | ||
| GenBank mRNA | BC018359.1 | - | 4 - 5 | 622 | 4341915 | 86 |
| D49357.1 | - | 4 - 5 | 418 | 86 | ||
| X69078.1 | - | 4 - 5 | 471 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001462.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfur amino acid metabolic process
selenium compound metabolic process
cellular amino acid metabolic process
S-adenosylmethionine biosynthetic process
one-carbon metabolic process
methylation
methionine adenosyltransferase activity
ATP binding
metal ion binding
selenomethionine adenosyltransferase activity
sulfur amino acid metabolic process
selenium compound metabolic process
cellular amino acid metabolic process
S-adenosylmethionine biosynthetic process
one-carbon metabolic process
methylation
methionine adenosyltransferase activity
ATP binding
metal ion binding
selenomethionine adenosyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018586.hg.1 | ENST00000372206.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018586.hg.1 | ENST00000372213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018586.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018586.hg.1 | TC10001462.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176449.hg.1 | TC1000011213.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |

MAT1A
Genomic Map