Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000429.2 | NP_000420.1 | 6 - 7 | 1026 | 80 | |
| XM_005269842.4 | XP_005269899.1 | 7 - 8 | 1256 | 80 | ||
| XM_005269843.4 | XP_005269900.1 | 6 - 7 | 1145 | 80 | ||
| GenBank mRNA | AK290820.1 | - | 6 - 7 | 892 | 80 | |
| BC018359.1 | - | 6 - 7 | 993 | 4341915 | 80 | |
| X69078.1 | - | 6 - 7 | 842 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001462.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfur amino acid metabolic process
selenium compound metabolic process
cellular amino acid metabolic process
S-adenosylmethionine biosynthetic process
one-carbon metabolic process
methylation
methionine adenosyltransferase activity
ATP binding
metal ion binding
selenomethionine adenosyltransferase activity
sulfur amino acid metabolic process
selenium compound metabolic process
cellular amino acid metabolic process
S-adenosylmethionine biosynthetic process
one-carbon metabolic process
methylation
methionine adenosyltransferase activity
ATP binding
metal ion binding
selenomethionine adenosyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018582.hg.1 | ENST00000372206.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018582.hg.1 | ENST00000372213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018582.hg.1 | ENST00000485270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018582.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018582.hg.1 | TC10001462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000455001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011213.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000372213.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000480845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | ENST00000485270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | NM_000429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176445.hg.1 | TC1000011213.hg.1 | Clariom D TC |

MAT1A
Genomic Map