| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017338.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017338.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017338.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017338.hg.1 |
TC10001369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174420.hg.1 |
TC1000010930.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000174423.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174423.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174423.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174423.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174423.hg.1 |
TC1000010930.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000299299.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
ENST00000493961.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_000281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010930.hg.1 |
NM_001289797.1 |
Refseq |