Assay Details
| Gene Symbol: ADAMTS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080722.4 | NP_542453.2 | 20 - 21 | 3368 | 96 | |
| NM_139155.3 | NP_631894.2 | 20 - 21 | 3377 | 96 | ||
| XM_011539300.2 | XP_011537602.1 | 17 - 18 | 2851 | 96 | ||
| XM_011539301.2 | XP_011537603.1 | 17 - 18 | 2374 | 96 | ||
| XM_011539302.2 | XP_011537604.1 | 17 - 18 | 2251 | 96 | ||
| XM_011539309.1 | XP_011537611.1 | 13 - 14 | 1961 | 96 | ||
| GenBank mRNA | AF358666.1 | - | 20 - 21 | 3071 | 96 | |
| AF366351.1 | - | 19 - 20 | 2998 | 96 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |

ADAMTS14
Genomic Map