Assay Details
| Gene Symbol: ADAMTS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080722.4 | NP_542453.2 | 8 - 9 | 1648 | 102 | |
| NM_139155.3 | NP_631894.2 | 8 - 9 | 1657 | 102 | ||
| XM_011539300.2 | XP_011537602.1 | 8 - 9 | 1641 | 102 | ||
| XM_011539301.2 | XP_011537603.1 | 5 - 6 | 654 | 102 | ||
| XM_011539302.2 | XP_011537604.1 | 5 - 6 | 531 | 102 | ||
| XM_011539303.2 | XP_011537605.1 | 8 - 9 | 1645 | 102 | ||
| XM_011539305.2 | XP_011537607.1 | 8 - 9 | 1645 | 102 | ||
| XM_011539306.2 | XP_011537608.1 | 8 - 9 | 1645 | 102 | ||
| XM_011539307.2 | XP_011537609.1 | 8 - 9 | 1645 | 102 | ||
| XM_011539308.2 | XP_011537610.1 | 8 - 9 | 1645 | 102 | ||
| GenBank mRNA | AF358666.1 | - | 8 - 9 | 1351 | 102 | |
| AF366351.1 | - | 7 - 8 | 1278 | 102 | ||
| AJ345098.1 | - | 8 - 9 | 1351 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000433.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005066.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005066.hg.1 | ENST00000373208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005066.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005066.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005066.hg.1 | TC10000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005067.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005067.hg.1 | ENST00000373208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005067.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005067.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005067.hg.1 | TC10000433.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007934.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153830.hg.1 | TC1000007934.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | ENST00000373207.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | ENST00000373208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | NM_080722.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | NM_139155.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153829.hg.1 | TC1000007934.hg.1 | Clariom D TC |

ADAMTS14
Genomic Map