Assay Details
| Gene Symbol: NOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014062.2 | NP_054781.1 | 2 - 3 | 251 | 101 | |
| GenBank mRNA | AB026125.1 | - | 2 - 3 | 209 | 101 | |
| AF190161.1 | - | 2 - 3 | 179 | 101 | ||
| AK001028.1 | - | 2 - 3 | 207 | 101 | ||
| AK314307.1 | - | 2 - 3 | 206 | 101 | ||
| AY487344.1 | - | 2 - 3 | 225 | 101 | ||
| BC064630.1 | - | 2 - 3 | 194 | 5740890 | 101 | |
| BM827373.1 | - | 2 - 3 | 227 | 101 | ||
| DB048531.1 | - | 2 - 3 | 251 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001226.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF190161.1,AK001028.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016952.hg.1 | ENST00000268802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016952.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016952.hg.1 | TC16001226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182930.hg.1 | NM_014062.2 | Refseq |

NOB1
Genomic Map