Assay Details
| Gene Symbol: NOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014062.2 | NP_054781.1 | 5 - 6 | 583 | 70 | |
| NR_074074.1 | 4 - 5 | 452 | 70 | |||
| GenBank mRNA | AB026125.1 | - | 5 - 6 | 541 | 70 | |
| AF190161.1 | - | 5 - 6 | 511 | 70 | ||
| AK001028.1 | - | 5 - 6 | 538 | 70 | ||
| AK314307.1 | - | 5 - 6 | 538 | 70 | ||
| AY487344.1 | - | 5 - 6 | 557 | 70 | ||
| BC000050.2 | - | 2 - 3 | 170 | 70 | ||
| BC064630.1 | - | 5 - 6 | 526 | 5740890 | 70 | |
| BG386829.1 | - | 2 | 396 | 70 | ||
| DA004856.1 | - | 4 - 5 | 408 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001226.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016949.hg.1 | ENST00000268802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016949.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016949.hg.1 | TC16001226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182924.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182923.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |

NOB1
Genomic Map