Assay Details
| Gene Symbol: NOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014062.2 | NP_054781.1 | 8 - 9 | 1030 | 99 | |
| NR_074074.1 | 7 - 8 | 899 | 99 | |||
| GenBank mRNA | AB026125.1 | - | 8 - 9 | 988 | 99 | |
| AF190161.1 | - | 8 - 9 | 958 | 99 | ||
| AK001028.1 | - | 8 - 9 | 985 | 99 | ||
| AK314307.1 | - | 8 - 9 | 985 | 99 | ||
| AY487344.1 | - | 8 - 9 | 1004 | 99 | ||
| BC000050.2 | - | 5 - 6 | 617 | 99 | ||
| BC064630.1 | - | 8 - 9 | 973 | 5740890 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001226.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008237.hg.1 | ENST00000575838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008237.hg.1 | ENST00000575838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016946.hg.1 | ENST00000268802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016946.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016946.hg.1 | TC16001226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182915.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182916.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182917.hg.1 | TC1600010734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008237.hg.1 | ENST00000575838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008237.hg.1 | ENST00000575838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | ENST00000268802.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | ENST00000569871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | NM_014062.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | NR_074074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182918.hg.1 | TC1600010734.hg.1 | Clariom D TC |

NOB1
Genomic Map