Assay Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152319.3 | NP_689532.1 | 4 - 5 | 267 | 100 | |
| XM_005268636.3 | XP_005268693.1 | 3 - 4 | 149 | 100 | ||
| XM_011537896.2 | XP_011536198.1 | 6 - 7 | 915 | 100 | ||
| XM_017018796.1 | XP_016874285.1 | 8 - 9 | 1105 | 100 | ||
| GenBank mRNA | AY255793.1 | - | 3 - 4 | 172 | 100 | |
| BC031670.2 | - | 3 - 4 | 193 | 5165130 | 100 | |
| BQ429029.1 | - | 4 - 5 | 270 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000351.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005268636.3,XM_011537896.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12004395.hg.1 | ENST00000314014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004395.hg.1 | ENST00000548364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004395.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004395.hg.1 | TC12000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152868.hg.1 | TC1200007522.hg.1 | Clariom D TC |

C12orf54
Genomic Map