Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017994.4 | NP_060464.1 | 2 - 3 | 422 | 90 | |
| XM_005250482.3 | XP_005250539.1 | 2 - 3 | 274 | 90 | ||
| GenBank mRNA | AK000961.1 | - | 2 - 3 | 408 | 90 | |
| AK222774.1 | - | 2 - 3 | 422 | 90 | ||
| BC008675.1 | - | 2 - 3 | 402 | 3853834 | 90 | |
| BC010519.1 | - | 2 - 3 | 376 | 4150551 | 90 | |
| BC012562.1 | - | 2 - 3 | 417 | 4332094 | 90 | |
| BC041764.2 | - | 2 - 3 | 391 | 4152659 | 90 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |

TMEM248
Genomic Map