Assay Details
| Gene Symbol: MRI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031727.2 | NP_001026897.1 | 4 - 5 | 795 | 93 | |
| NM_032285.2 | NP_115661.1 | 4 - 5 | 654 | 93 | ||
| XM_011528355.2 | XP_011526657.1 | 4 - 5 | 730 | 93 | ||
| XM_011528356.2 | XP_011526658.1 | 4 - 5 | 818 | 93 | ||
| GenBank mRNA | AL834276.1 | - | 4 - 5 | 645 | 93 | |
| AY358176.1 | - | 4 - 5 | 772 | 93 | ||
| BC001703.1 | - | 4 - 5 | 769 | 3501899 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000239.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19003535.hg.1 | ENST00000319545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003535.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003535.hg.1 | TC19000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003538.hg.1 | ENST00000040663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003538.hg.1 | ENST00000319545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003538.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003538.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003538.hg.1 | TC19000239.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | TC1900011674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199553.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | TC1900011674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199544.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000588526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000589290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | ENST00000593245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011674.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | TC1900011674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199552.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | NM_032285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | TC1900011674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000040663.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000319545.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | ENST00000591688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | NM_001031727.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199550.hg.1 | NM_032285.3 | Refseq |

MRI1
Genomic Map