Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146319.1 | NP_001139791.1 | 2 - 3 | 486 | 67 | |
| NM_001146322.1 | NP_001139794.1 | 2 - 3 | 297 | 67 | ||
| NM_023927.2 | NP_076416.2 | 2 - 3 | 635 | 67 | ||
| XM_005272057.3 | XP_005272114.1 | 2 - 3 | 373 | 67 | ||
| XM_005272058.3 | XP_005272115.1 | 2 - 3 | 373 | 67 | ||
| XM_005272060.3 | XP_005272117.1 | 2 - 3 | 460 | 67 | ||
| XM_005272061.3 | XP_005272118.1 | 2 - 3 | 455 | 67 | ||
| XM_005272062.3 | XP_005272119.1 | 2 - 3 | 421 | 67 | ||
| XM_005272063.3 | XP_005272120.1 | 2 - 3 | 570 | 67 | ||
| XM_011543593.2 | XP_011541895.1 | 2 - 3 | 460 | 67 | ||
| XM_011543594.2 | XP_011541896.1 | 2 - 3 | 232 | 67 | ||
| GenBank mRNA | AK024966.1 | - | 2 - 3 | 403 | 67 | |
| AK225829.1 | - | 2 - 3 | 403 | 67 | ||
| AK298009.1 | - | 2 - 3 | 220 | 67 | ||
| AK298349.1 | - | 2 - 3 | 308 | 67 | ||
| AK300154.1 | - | 2 - 3 | 431 | 67 | ||
| AK303857.1 | - | 2 - 3 | 237 | 67 | ||
| AY116970.1 | - | 2 - 3 | 205 | 67 | ||
| BC008590.1 | - | 2 - 3 | 368 | 4148772 | 67 | |
| DC413835.1 | - | 2 - 3 | 251 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298009.1 transcripts.
Set Membership: |
|

GRAMD3
Genomic Map