Assay Details
| Gene Symbol: PNPLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025225.2 | NP_079501.2 | 1 - 2 | 360 | 111 | |
| GenBank mRNA | AK025665.1 | - | 1 - 2 | 360 | 111 | |
| AK315166.1 | - | 1 - 2 | 294 | 111 | ||
| AL138578.2 | - | 1 - 2 | 360 | 111 | ||
| BC065195.1 | - | 1 - 2 | 288 | 5243623 | 111 | |
| CR456476.1 | - | 1 - 2 | 188 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001477.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
triglyceride biosynthetic process
triglyceride catabolic process
triglyceride acyl-chain remodeling
acylglycerol acyl-chain remodeling
lipid homeostasis
lipoprotein lipase activity
phospholipase A2 activity
triglyceride lipase activity
acylglycerol O-acyltransferase activity
mono-olein transacylation activity
diolein transacylation activity
triglyceride biosynthetic process
triglyceride catabolic process
triglyceride acyl-chain remodeling
acylglycerol acyl-chain remodeling
lipid homeostasis
lipoprotein lipase activity
phospholipase A2 activity
triglyceride lipase activity
acylglycerol O-acyltransferase activity
mono-olein transacylation activity
diolein transacylation activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22006996.hg.1 | ENST00000216180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006996.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006996.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006996.hg.1 | TC22001477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007000.hg.1 | ENST00000216180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007000.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007000.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007000.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007000.hg.1 | TC22001477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006993.hg.1 | ENST00000216180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006993.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006993.hg.1 | TC22001477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169198.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169198.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169198.hg.1 | TC2200009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169198.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169198.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | ENST00000497129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009281.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | TC2200009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169200.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | TC2200009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169201.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | NM_025225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | TC2200009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000216180.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000423180.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | ENST00000478713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169204.hg.1 | NM_025225.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 197774 | - | - |

PNPLA3
Genomic Map