| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007005.hg.1 |
ENST00000216180.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007005.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007005.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007005.hg.1 |
TC22001477.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
TC2200009281.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169209.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000216180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000423180.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000478713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
ENST00000497129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009281.hg.1 |
NM_025225.2 |
Refseq |