Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
TC03000770.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
ENST00000273482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
ENST00000416210.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
ENST00000476941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
NM_001251845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014068.hg.1 |
NM_003304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000273482.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000460401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000476941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000612385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
NM_001251845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
NM_003304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000273482.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000460401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000476941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
ENST00000612385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
NM_001251845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
NM_003304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164813.hg.1 |
TC0300009035.hg.1 |
Clariom D TC |