Assay Details
| Gene Symbol: TTC39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080494.3 | NP_001073963.1 | 15 - 16 | 1489 | 98 | |
| NM_001144832.2 | NP_001138304.1 | 15 - 16 | 1464 | 98 | ||
| NM_001297663.1 | NP_001284592.1 | 15 - 16 | 1466 | 98 | ||
| NM_001297664.1 | NP_001284593.1 | 15 - 16 | 1502 | 98 | ||
| NM_001297665.1 | NP_001284594.1 | 15 - 16 | 1594 | 98 | ||
| NM_001297666.1 | NP_001284595.1 | 4 - 5 | 497 | 98 | ||
| NM_001297667.1 | NP_001284596.1 | 3 - 4 | 405 | 98 | ||
| XM_005270644.2 | XP_005270701.1 | 15 - 16 | 1495 | 98 | ||
| XM_006710471.2 | XP_006710534.1 | 15 - 16 | 1592 | 98 | ||
| XM_011541048.2 | XP_011539350.1 | 15 - 16 | 1569 | 98 | ||
| XM_011541049.2 | XP_011539351.1 | 15 - 16 | 1708 | 98 | ||
| XM_011541050.2 | XP_011539352.1 | 15 - 16 | 1569 | 98 | ||
| XM_011541051.2 | XP_011539353.1 | 15 - 16 | 1602 | 98 | ||
| XM_011541052.2 | XP_011539354.1 | 15 - 16 | 1548 | 98 | ||
| XM_011541053.2 | XP_011539355.1 | 15 - 16 | 1586 | 98 | ||
| XM_011541054.2 | XP_011539356.1 | 15 - 16 | 1422 | 98 | ||
| XM_017000715.1 | XP_016856204.1 | 15 - 16 | 1469 | 98 | ||
| GenBank mRNA | AF007170.1 | - | 15 - 16 | 1370 | 98 | |
| AK298019.1 | - | 15 - 16 | 1473 | 98 | ||
| AK301587.1 | - | 15 - 16 | 1462 | 98 | ||
| BC004399.1 | - | 4 - 5 | 458 | 98 | ||
| BC028374.2 | - | 15 - 16 | 1536 | 98 | ||
| BQ227134.1 | - | 3 - 4 | 416 | 98 | ||
More Information
Set Membership: |
|

TTC39A
Genomic Map