Assay Details
| Gene Symbol: SOX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007017.2 | NP_008948.1 | 3 - 4 | 1727 | 61 | |
| GenBank mRNA | AB022441.1 | - | 3 - 4 | 1680 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012605.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187973.hg.1 | TC0500012605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187974.hg.1 | TC0500012605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187977.hg.1 | TC0500012605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187975.hg.1 | TC0500012605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000265007.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000311371.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | ENST00000519442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_001308165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_007017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | NM_178424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187976.hg.1 | TC0500012605.hg.1 | Clariom D TC |

SOX30
Genomic Map