Assay Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282607.1 | NP_001269536.1 | 3 - 4 | 429 | 86 | |
| NM_144967.3 | NP_659404.2 | 3 - 4 | 665 | 86 | ||
| XM_006724729.1 | XP_006724792.1 | 2 - 3 | 136 | 86 | ||
| XM_011531280.1 | XP_011529582.1 | 2 - 3 | 153 | 86 | ||
| GenBank mRNA | AK128203.1 | - | 3 - 4 | 665 | 86 | |
| AK294274.1 | - | 3 - 4 | 388 | 86 | ||
| AK296748.1 | - | 3 - 4 | 407 | 86 | ||
| BC063790.1 | - | 3 - 4 | 264 | 6196091 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000625.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC063790.1,XM_006724729.1,XM_011531280.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | ENST00000276211.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | ENST00000370922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | ENST00000412432.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008276.hg.1 | TC0X000625.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008415.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000276211.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000370921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000370922.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000412432.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | ENST00000423277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | NM_001282607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | NM_144967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157329.hg.1 | TC0X00008415.hg.1 | Clariom D TC |

ARHGAP36
Genomic Map