Assay Details
| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048164.2 | NP_001041629.1 | 1 - 2 | 153 | 64 | |
| GenBank mRNA | AK075014.1 | - | 1 - 2 | 153 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001117.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK075014.1,NM_001048164.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transport
arginine transport
lysine transport
ornithine transport
regulation of TOR signaling
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
basic amino acid transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity
amino acid transport
arginine transport
lysine transport
ornithine transport
regulation of TOR signaling
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
basic amino acid transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015209.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015209.hg.1 | ENST00000374299.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015209.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015209.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015209.hg.1 | TC0X001117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168098.hg.1 | TC0X00009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |

SLC7A3
Genomic Map