Assay Details
| Gene Symbol: MBL1P | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002724.2 | 3 | 1243 | 99 | |||
| GenBank mRNA | AF019382.1 | - | LIT2047 MBL1P1 |
3 | 1242 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000574.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006216.hg.1 | TC10000574.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006213.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006213.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006213.hg.1 | TC10000574.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011179.hg.1 | ENST00000402234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155623.hg.1 | TC1000008216.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155624.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155624.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155624.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155624.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155624.hg.1 | TC1000008216.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000421889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000453174.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008216.hg.1 | NR_033857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155622.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155622.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155622.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155622.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155622.hg.1 | TC1000008216.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | ENST00000480805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | ENST00000497731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | NR_002724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155632.hg.1 | TC1000008216.hg.1 | Clariom D TC |

MBL1P
Genomic Map