| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000357054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000375202.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000395220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000529487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000534123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
ENST00000541524.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
NM_020893.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
NR_036527.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
NR_036528.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
NR_036529.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005241.hg.1 |
TC09000489.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000357054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000375206.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000375206.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000532526.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000532526.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000534123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000534123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NM_020893.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036527.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036528.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036529.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000357054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000375206.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000375206.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000529787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000532526.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000532526.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000534123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
ENST00000534123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NM_020893.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036527.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036528.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
NR_036529.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900154590.hg.1 |
TC0900008145.hg.1 |
Clariom D TC |