Assay Details
| Gene Symbol: EPHX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142886.1 | NP_001136358.1 | 3 - 4 | 508 | 73 | |
| NM_024794.2 | NP_079070.1 | 2 - 3 | 544 | 73 | ||
| GenBank mRNA | AK026061.1 | - | 2 - 3 | 544 | 73 | |
| BC115002.1 | - | 3 - 4 | 418 | 7482856 | 73 | |
| BC132958.1 | - | 2 - 3 | 508 | 40146980 | 73 | |
| BC132960.1 | - | 2 - 3 | 508 | 40146982 | 73 | |
| DA378406.1 | - | 3 - 4 | 508 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001251.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017580.hg.1 | ENST00000221730.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017580.hg.1 | ENST00000435261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017580.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017580.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017580.hg.1 | TC19001251.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | TC1900009858.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180793.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000594042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009858.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | NM_024794.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | TC1900009858.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000221730.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000435261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | ENST00000602233.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | NM_001142886.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180792.hg.1 | NM_024794.2 | Refseq |

EPHX3
Genomic Map