Assay Details
Gene Symbol: TXNRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282512.1 | NP_001269441.1 | 8 - 9 | 854 | 78 | |
NM_006440.4 | NP_006431.2 | 8 - 9 | 854 | 78 | ||
GenBank mRNA | AB019694.1 | - | 8 - 9 | 665 | 78 | |
AB019695.1 | - | 8 - 9 | 759 | 78 | ||
AB209724.1 | - | 8 - 9 | 632 | 78 | ||
AF044212.1 | - | 8 - 9 | 675 | 78 | ||
AF106697.1 | - | 8 - 9 | 854 | 78 | ||
AF166126.1 | - | 3 - 4 | 739 | 78 | ||
AF166127.1 | - | 8 - 9 | 759 | 78 | ||
AF171054.1 | - | 8 - 9 | 672 | 78 | ||
AF201385.1 | - | 8 - 9 | 662 | 78 | ||
BC007489.2 | - | 7 - 8 | 549 | 78 | ||
BC099923.1 | - | 8 - 9 | 666 | 5213425 | 78 | |
BC117354.1 | - | 8 - 9 | 634 | 40125905 | 78 | |
BX957216.1 | - | 8 - 9 | 656 | 78 | ||
CR749428.1 | - | 8 - 9 | 656 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF166126.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
