Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000275.2 | NP_000266.2 | 20 - 21 | 2251 | 111 | |
| NM_001300984.1 | NP_001287913.1 | 19 - 20 | 2179 | 111 | ||
| XM_011521640.2 | XP_011519942.1 | 20 - 21 | 2281 | 111 | ||
| XM_017022255.1 | XP_016877744.1 | 20 - 21 | 2232 | 111 | ||
| XM_017022256.1 | XP_016877745.1 | 20 - 21 | 2729 | 111 | ||
| XM_017022257.1 | XP_016877746.1 | 19 - 20 | 2182 | 111 | ||
| XM_017022258.1 | XP_016877747.1 | 20 - 21 | 2254 | 111 | ||
| XM_017022259.1 | XP_016877748.1 | 19 - 20 | 2182 | 111 | ||
| XM_017022260.1 | XP_016877749.1 | 18 - 19 | 2116 | 111 | ||
| XM_017022261.1 | XP_016877750.1 | 19 - 20 | 2748 | 111 | ||
| XM_017022262.1 | XP_016877751.1 | 20 - 21 | 2254 | 111 | ||
| GenBank mRNA | BC012097.1 | - | 19 - 20 | 2179 | 4641135 | 111 |
| M99564.1 | - | 20 - 21 | 2193 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001091.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009867.hg.1 | ENST00000353809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009867.hg.1 | ENST00000354638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009867.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009867.hg.1 | TC15001091.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009868.hg.1 | ENST00000353809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009868.hg.1 | ENST00000354638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009868.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009868.hg.1 | TC15001091.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163483.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163482.hg.1 | NM_000275.2 | Refseq |

OCA2
Genomic Map