Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000275.2 | NP_000266.2 | 8 - 9 | 995 | 130 | |
| NM_001300984.1 | NP_001287913.1 | 8 - 9 | 995 | 130 | ||
| XM_011521640.2 | XP_011519942.1 | 8 - 9 | 1025 | 130 | ||
| XM_017022255.1 | XP_016877744.1 | 8 - 9 | 976 | 130 | ||
| XM_017022256.1 | XP_016877745.1 | 8 - 9 | 1473 | 130 | ||
| XM_017022257.1 | XP_016877746.1 | 8 - 9 | 998 | 130 | ||
| XM_017022258.1 | XP_016877747.1 | 8 - 9 | 998 | 130 | ||
| XM_017022259.1 | XP_016877748.1 | 8 - 9 | 998 | 130 | ||
| XM_017022260.1 | XP_016877749.1 | 8 - 9 | 998 | 130 | ||
| XM_017022261.1 | XP_016877750.1 | 7 - 8 | 1492 | 130 | ||
| XM_017022262.1 | XP_016877751.1 | 8 - 9 | 998 | 130 | ||
| XM_017022263.1 | XP_016877752.1 | 8 - 9 | 998 | 130 | ||
| XM_017022264.1 | XP_016877753.1 | 8 - 9 | 998 | 130 | ||
| XM_017022265.1 | XP_016877754.1 | 8 - 9 | 998 | 130 | ||
| GenBank mRNA | BC012097.1 | - | 8 - 9 | 995 | 4641135 | 130 |
| M99564.1 | - | 8 - 9 | 937 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001091.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009883.hg.1 | ENST00000353809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009883.hg.1 | ENST00000354638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009883.hg.1 | ENST00000382996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009883.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009883.hg.1 | TC15001091.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163499.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163501.hg.1 | NM_000275.2 | Refseq |

OCA2
Genomic Map